Learn More
Med Vet International Miltex Halsted Mosquito Hemostatic Forceps, 5in., Curved, Extra Delicate
Supplier:  Med Vet International MIL710
Encompass_Preferred
The Miltex 7-10 Halsted mosquito forceps are made of OR grade German stainless steel and measure 5" in length. They have a finger ring handle with a ratchet lock and curved tips that are serrated and extra delicate.
Catalog No. 50-283-0744