Learn More
Med Vet International Miltex Kantrowitz Thoracic Forceps, 7.75in, Delicate Right Angle Jaws, German, 25-836
Supplier: Med Vet International MIL25836
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Kantrowitz Thoracic Forceps. Kantrowitz Thoracic Forceps can be used during thoracic surgery. They are made from premium-grade German stainless steel. Specifications: 7.75in, Delicate Right Angle Jaws, 25-836