Learn More
Med Vet International Miltex Kelly Placenta Forceps, 12.5in, Curved, Serrated Ring Jaws, German, 30-2505
Supplier:  Med Vet International MIL302505
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Kelly Placenta Forceps. Kelly Placenta Forceps are commonly used post-childbirth to aid the mother in pushing out the placenta. They are made from premium-grade German stainless steel. Specifications: 12.5in, Curved, Serrated Ring Jaws, 30-2505
Catalog No. 50-283-2427