Learn More
Med Vet International Miltex Mcpherson Iris Suturing Forceps, 3.5in, Angled, Tying Platform, 0.3mm Wide At Tip, German, 18-949
Supplier:  Med Vet International MIL18949
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Mcpherson Iris Suturing Forceps. McPherson Iris Suturing Forceps are widely used for suturing in ophthalmic procedures. They are made from premium-grade German stainless steel. Specifications: 3.5in, Angled, Tying Platform, 0.3mm Wide At Tip, 18-949
Catalog No. 50-283-1812