Learn More
Med Vet International Miltex Meisterhand Rochester-Carmalt Forceps, 6.5in, Straight, German, Mh7-170
Supplier:  Med Vet International MILMH7170
Encompass_Preferred
The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Rochester-Carmalt Forceps. Nicknamed the "stars and stripes hemostat, " the Rochester-Carmalt Forceps are characterized by the longitudinal serrations that run the length of the blade with cross-hatching at the tip. These large, crushing hemostatic forceps are a choice instrument for clamping blood vessels and large tissues or ligating pedicles. They are made from premium-grade German stainless steel. Specifications: 6.5in, Straight, MH7-170
Catalog No. 50-283-2163