Learn More
Med Vet International Miltex Swiss Cilia And Suture Forceps, 3.75in, 2.8mm Wide Slanted Smooth Jaws, 18-1107
Supplier:  Med Vet International MIL181107
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Swiss Cilia And Suture Forceps. Swiss Cilia Forceps are a multipurpose instrument for ophthalmologic procedures involving the eyelid. They are made from premium-grade surgical stainless steel. Specifications: 3.75in, 2.8mm Wide Slanted Smooth Jaws, 18-1107
Catalog No. 50-264-3556