Learn More
Med Vet International Hartmann Mosquito Forceps, 3.5in, Curved, German Grade, Von Klaus
Supplier:  Med Vet International VK1153309
Encompass_Preferred
3.5in. Hartmann Mosquito Forceps is a German grade surgical instrument and are typically used as both a hemostat and fine tissue dissection. They are curved and are typically shorter and heavier than Halstead Mosquito Forceps. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1906