Learn More
Med Vet International Rochester-Pean Forceps, 6.25", Curved, German Grade, Von Klaus
Supplier:  Med Vet International VK1161116
Encompass_Preferred
6.25in. Rochester-Pean Forceps is a German grade surgical instrument and are used for clamping larger tissue and vessels. They are curved with serrated jaws. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1913