Learn More
World Precision Instrument Round Hollow Handled Forceps, 15cm, Straight
SDP
Supplier:  World Precision Instrument 14113
Encompass
Hollowing out the handles of long, round-handled instruments decreases the overall weight. This makes them efficient and non-fatiguing when operating under a microscope. Made from high quality stainless steel, these instruments are an excellent addition to a microsurgical instrument kit.
Catalog No. 50-822-219