Learn More
Med Vet International Miltex Padgett Crow Coagulating Forceps, 9.25in, Tungsten Carbide, insulated Handle, German, PM-9043
Supplier:  Med Vet International MILPM9043
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Padgett Crow Coagulating Forceps. Crow Coagulating Forceps are for monopolar coagulation. They are made from premium-grade German stainless steel. Specifications: 9.25in, Tungsten Carbide, insulated Handle, PM-9043
Catalog No. 50-264-3478