Learn More
Med Vet International Miltex Eye Dressing Forceps, 4in., Straight, Standard Pattern, 0.8mm Wide Serrated Tips
Supplier:  Med Vet International MIL18780
Encompass_Preferred
The Miltex 18-780 eye dressing forceps are made of OR grade German stainless steel and measure 4" in length. They have a non-locking thumb handle and straight, 0.8mm wide serrated tips.
Catalog No. 50-283-0524