Learn More
Med Vet International Miltex Vantage Hartman Mosquito Hemostatic Forceps, 3.5in, Straight
Supplier:  Med Vet International MILV9724
Encompass_Preferred
The Miltex Vantage V97-24 Hartman mosquito forceps are made from high quality Japanese stainless steel and measure 3.5" in length. They have a finger ring handle with a ratchet lock and straight tips that are serrated.
Catalog No. 50-252-2265