Learn More
World Precision Instrument Kelly Hemostatic Forceps, 14cm, Straight
SDP
Supplier:  World Precision Instrument 501241
Encompass
Kelly hemostatic forceps are used to clamp blood vessels and hold heavy tissues in place. Kelly forceps, both straight and curved, are available. Our forceps feature a locking ratchet and wider finger rings for easy handling.
Catalog No. 50-822-589
 Restricted Item