Learn More
Roboz Surgical FORCEPS NO2 INOX 120MM LNTH
SDP
Supplier:  Roboz Surgical RS5041
Encompass
Dumont Tweezers; Pattern #2; .34 X .14mm Tip Size; 120mm Length; Material: Inox
Catalog No. NC9491283