Learn More
Research Products International Corp Ergonomic Forceps, Smooth, 1.5mm Tip, 7 Inches Long
SDP
Supplier:  Research Products International Corp 248831
Encompass
Ergonomic forceps are specially designed with a wide paddle handle and reduced tension. Lightweight forceps reduce hand fatigue and are effortless to operate. Forceps are easy to manage during procedures involving tissue dissection, embedding, preparation, and much more.
Catalog No. 50-212-611