Learn More
BURKLE INC Forceps, stainless steel, sharp, straight, 6.30 in. (160 mm)
Supplier:  BURKLE INC 53860204
Encompass_Preferred
The sturdy stainless steel forceps are ideal for use in laboratories. Pointed ends with a fluted internal structure ensure reliable handling of different samples. The fluted handle offers a good grip. The laboratory tweezers are suitable for use in precise work in chemistry, the pharmaceutical industry, cosmetics and food & beverage.
Catalog No. 50-308-1215