Learn More
BURKLE INC Forceps, stainless steel, sharp, bent form, 4.13 in. (105 mm)
Supplier:  BURKLE INC 53860100
Encompass_Preferred
The stainless steel forceps with angled shape allow for accurate work in the laboratory. The pointed, fluted ends can be used to work reliably with sensitive materials, tissue or samples. Flutings on the handle ensure a slip-free grip of the stainless steel tweezers. The angled laboratory tweezers are suitable for use in chemistry, the pharmaceutical industry, cosmetics and food & beverage.
Catalog No. 50-308-1391