Learn More
Med Vet International Desmarres Chalazion Forceps, 3.5in, Small, German Grade, Von Klaus
Supplier:  Med Vet International VK1605501
Encompass_Preferred
3.5in. Desmarres Chalazion forceps are German grade surgical instrument use in ophthalmic procedures. They have a small inside ring that is 17mm x 11mm. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1933