Learn More
Med Vet International 5.25-in Economy Kelly Forceps, Curved
Supplier:  Med Vet International 4521
Encompass_Preferred
Kelly hemostatic forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection.
Catalog No. 50-222-3996