Learn More
Med Vet International Kelly Forceps, 5.25 in, Straight, Economy
Supplier:  Med Vet International 4520
Encompass_Preferred
Kelly hemostatic forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection.
Catalog No. 50-222-3995