Learn More
Med Vet International 6.25" Economy Kelly Forceps, Straight
Supplier:  Med Vet International 4522
Encompass_Preferred
Kelly hemostatic forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle.
Catalog No. 50-222-3922