Learn More
Med Vet International 8in Rochster-Carmalt Forceps, Curved, German Grade, Von Klas
Supplier:  Med Vet International VK1163720
Encompass_Preferred
This 8' Rochester-Carmalt forceps is a German quality surgical instrument. It is curved with serrated tips. Rochester-Carmalt forceps are a type of hemostatic clamp that is used to stop blood flow of large vessels or to grip tough tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-1951