Learn More
Fine Science Tools Adson Forceps, DeBakey Serrations, 12.5 cm
SDP
Supplier:  Fine Science Tools 1161712
Encompass
Tip Shape: Straight/ Tips: DeBakey Serrations/ Tip Dimensions: 1.75 x 1.5mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm
Catalog No. NC9280122