Learn More
Grainger Fork End 316 SS Cable Size 3/8
Supplier:  Grainger 16X667
Encompass_Preferred
The Grainger 316 stainless steel fork end fittings for wire rope are designed to terminate wire rope for lifting and pulling applications. They are made from 316 stainless steel for corrosion resistance and have a threaded stud with nut for attaching to equipment. Fork openings accommodate wire rope diameters from 1/4" to 1-1/4". Load ratings vary by size from 1,250 lbs to 23,000 lbs. Fork end fittings allow wire rope to be easily attached to cranes, hoists, rigging and lifting equipment. Grainger offers a range of sizes to fit different wire rope diameters and load requirements.
Catalog No. 50-276-5072