Learn More
Grainger Fork End 316 SS Cable Size 5/16
Supplier:  Grainger 16X666
Encompass_Preferred
The Grainger 316 stainless steel fork end fittings for wire rope are designed for use with wire rope to create an eye or loop. They are made from 316 stainless steel for corrosion resistance and feature a threaded stud with nut to secure the wire rope. Fork openings range from 5/16 to 1 inch to accommodate different wire rope diameters. Load ratings vary by size from 1,250 lbs to 9,100 lbs. Grainger offers a wide selection of stainless steel fork end fittings for wire rope in multiple sizes to meet application needs.
Catalog No. 50-276-5071