Learn More
Polysciences, Inc. Formaldehyde, 10%, Methanol free, Ultra Pure, Suitable for both Electron and Light Microscopy
SDP
Supplier:  Polysciences, Inc. 040184
Encompass_Preferred
Formaldehyde, 10%, Methanol free, Ultra Pure, Suitable for both Electron and Light Microscopy, Irritant, Lachrymator, Expected carcinogen, Toxic (Ingestion), Store at room temperature, Boiling Point: >200degreeF, Melting Point: 5degreeF
Catalog No. NC9200219
 Restricted Item