Learn More
Chem Service Inc FORMALDEHYDE DNPH DERIV 100MG
SDP
Supplier:  Chem Service Inc N12011100MG
Encompass_Preferred
Formaldehyde (DNPH Derivative); Solid; 100mg; M.W. 210.17; C7H6N4O4; M.P. 153 deg. to 156 deg. C; CAS: 1081-15-8; Stored in a cool dry place, Stored only with compatible chemicals
Catalog No. NC9201772