Learn More
Electron Microscopy Sciences Formaldehyde Glutaraldehyde 3% Sodium Cacodylate Buffer, 7.4 pH 10 ML
SDP
Supplier:  Electron Microscopy Sciences 15950
Encompass_Preferred
Premixed, prebuffered fixatives, are pH adjusted for immediate laboratory use. They are freshly prepared upon receipt of order to ensure maximum shelf life. The fixatives are filled in 20 ml glass vials (10 ml/vial). This allows for the immediate fixation of your specimens without costly and time consuming transfer associated with ampoule contained fixatives. In Sodium Cacodylate.
pH: 7.4
Catalog No. 50-259-42