Learn More
Electron Microscopy Sciences Formalin 10% Buffered in Acetate Buffer, 6.0 to 7.1 pH at 20°C pH 1 LT
SDP
Supplier:  Electron Microscopy Sciences 1573801
Encompass_Preferred
Buffered with Sodium Acetate in accordance with the A.F.I.P. manual of Histology. Filtered and ready-to-use. pH 6.0 to ph 7.1 at 20 degree C
pH: 6.0 to 7.1 pH at 20°C
Catalog No. 50-980-498