Learn More
Electron Microscopy Sciences Formalin Solution 50% 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2669803
Encompass_Preferred
Used in Hirano-Zimmerman Method for Nerve Cells and Fibers for staining of nerofibrils, dendrites, axis cylinders, senile plaques, etc. See AFIP Manual of Histological Staining Methods, 3rd ed., Ed. L.Luna: New York: McGraw-Hill Pub., c 1968, p. 198 or Hirano, A. and Zimmerman, H.M., Arch. Neurol., 6:114-122, 1962
Catalog No. 50-318-92