Learn More
IBI SCIENTIFIC 3P Spectral Grade Formamide, 450ml
Supplier:  IBI SCIENTIFIC 3P IB72024
Encompass_Preferred
Spectral Grade Formamide is used to denature DNA and RNA in hybridization and electrophoresis applications via separating the double helix into two strands. It's supplied distilled, deionized and packaged under nitrogen. CAS#: 75-12-7.
Catalog No. NC9710732