Learn More
Medchemexpress LLC Formamide-d3 | 43380-64-9 | MFCD00144375 | 99.7% | 48.06 g/mol | CD3NO | 50mg
SDP
Supplier:  Medchemexpress LLC HYY0842S150MG
Encompass_Preferred
Formamide-d3 is the deuterium labeled Formamide (HY-Y0842) Formamide is an effective DNA denaturant that significantly reduces the stability of DNA in buffer solutions Formamide can also be used as a decalcifying agent for rat cardiac cells Additionally formamide is widely used as a solvent or chemical raw material for ion compounds resins and plasticizers [1][2][3][4][5][6][7]
Catalog No. 50-002-68529
 Restricted Item