Learn More
MilliporeSigma Formamide, Deionized, 500ml
Supplier:  MilliporeSigma S4117
Encompass
This product is intended for use in hybridization applications for in situ, Southern, Northern, dot and slot blot analyses, and colony, plaque, and YAC screening. Formamide is used primarily as a component in probe hybridization solutions to promote probe hybridization specificity. It denatures nucleic acids by disrupting hydrogen bonds. 500ml
Catalog No. NC9569627