Learn More
TARGETMOL CHEMICALS INC Formiminoglutamic acid 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3185125MG
Encompass_Preferred
Also available in 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Formiminoglutamic acid (NSC 334078) is a biomarker for formylaminoglutaminuria, an indicator of folate deficiency. Purity 98.07%
Catalog No. 50-287-7799