Learn More
Medchemexpress LLC Formiminoglutamic acid | 816-90-0 | ≥99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1293981MG
Encompass_Preferred
Formiminoglutamic acid is an endogenous metabolite that may serve as an index of folic-acid deficiency. This product is intended for research purposes only.
  • Endogenous metabolite
  • Potential index of folic-acid deficiency
  • For research use only
Catalog No. 50-003-40369