Learn More
Medchemexpress LLC Formiminoglutamic acid | 816-90-0 | MFCD00037305 | ≥99.0% | 174.15 g/mol | C6H10N2O4 | 5 MG

Supplier: Medchemexpress LLC HY1293985MG
Formiminoglutamic acid is an endogenous metabolite used in biochemical and clinical research as an indicator of folic-acid (folate) deficiency. The compound is supplied in purified form for use as a reference standard and reagent in metabolic studies and analytical testing.
- Endogenous metabolite used as an indicator of folate deficiency.
- Purity ≥99.0% suitable for analytical and reference use.
- Molecular weight 174.15 g/mol; formula C6H10N2O4.
- Available in small pack sizes (1 mg, 5 mg, 10 mg, 50 mg) for laboratory workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.