Learn More
TARGETMOL CHEMICALS INC FORMOSANIN C 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T35195MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Formosanin C (Polyphyllin B) is a compound isolated from Rhizoma Paridis showed pro-apoptosis and immunoregulation with antitumor activity in cultured cells and animal systems. purity: 99%
Catalog No. 50-258-0229