Learn More
Medchemexpress LLC Formoterol fumarate | 43229-80-7 | >99.9% | C19H24N2O4 . 1/2C4H4O4 | 100 MG
Supplier: Medchemexpress LLC HYB0010100MG
Formoterol fumarate is a selective β2-adrenoceptor agonist, at least as selective as Salbutamol and Terbutaline. It relaxes acetylcholine-pre-contracted human isolated segmental bronchi and can be used for research related to chronic obstructive pulmonary disease and bronchial asthma.
- Selective β2-adrenoceptor agonist
- Relaxes acetylcholine-pre-contracted human isolated segmental bronchi
- Suitable for research on chronic obstructive pulmonary disease
- Suitable for research on bronchial asthma