Learn More
Chem Service Inc Formothion, 2540-82-1, MFCD00055528, 100 mg
SDP
Supplier:  Chem Service Inc N12017100MG
Encompass_Preferred
Molecular Formula: C6-H12-N-O4-P-S2, Molecular Weight: 257.27, Classification: Insecticide, ORGANOPHOSPHOROUS PESTICIDE, ORGANOPHOSPHOROUS PESTICIDE, Alternate Name(s): 2-DIMETHOXYPHOSPHINOTHIOYLTHIO-N-FORMYL-N, -METHYLACETAMIDE, ANTHIO.
Catalog No. NC1550630