Learn More
Ted Pella Inc Formvar/Carbon 200 mesh, Copper 100/pk
SDP
Supplier:  Ted Pella Inc 01801
Encompass
A Formvar film covered with a "light" layer of carbon. The heat and electrical conducting properties of carbon help to stabilize the Formvar films when exposed to the electron beam. This is a resilient, all-purpose specimen support film, ideal for mounting thin sections and for applications using lower ranges of magnification.
Catalog No. NC0258037