Learn More
Medchemexpress LLC Forodesine hydrochloride | 284490-13-7 | 98.4% | 302.71 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16209
Encompass_Preferred
Forodesine hydrochloride is a highly potent and orally active purine nucleoside phosphorylase (PNP) inhibitor with IC50 values ranging from 0.48 to 1.57 nM for human, mouse, rat, monkey and dog PNP. It is a potent human lymphocyte proliferation inhibitor and can induce apoptosis in leukemic cells by increasing dGTP levels.
  • Potent and orally active purine nucleoside phosphorylase (PNP) inhibitor.
  • Inhibits human lymphocyte proliferation.
  • Induces apoptosis in leukemic cells.
  • Excellent oral bioavailability in mice.
  • Effective in prolonging life span in mouse models.
Catalog No. 50-003-91398