Learn More
Medchemexpress LLC Forodesine hydrochloride | 284490-13-7 | 99.9% | 302.71 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1620910MM/1ML
Encompass_Preferred
Forodesine hydrochloride is a highly potent and orally active purine nucleoside phosphorylase (PNP) inhibitor. It acts as a potent human lymphocyte proliferation inhibitor and can induce apoptosis in leukemic cells by increasing dGTP levels. It has excellent oral bioavailability in mice.
  • Highly potent and orally active purine nucleoside phosphorylase (PNP) inhibitor
  • Potent human lymphocyte proliferation inhibitor
  • Induces apoptosis in leukemic cells
  • Excellent oral bioavailability in mice
Catalog No. 50-003-92234