Learn More
Medchemexpress LLC Foropafant | 136468-36-5 | 100.0% | 100 MG
Supplier: Medchemexpress LLC HY128694100MG
Foropafant is a highly potent, competitive, selective, and orally active antagonist of the platelet-activating factor (PAF) receptor. It is characterized by a Ki value of 57 pM for [3H]PAF binding, which is significantly lower than unlabeled PAF, and effectively inhibits PAF-induced aggregation in both rabbit and human platelets. This product is intended for research use only and should be handled by qualified personnel.
- Highly potent and selective PAF receptor antagonist
- Orally active compound
- Inhibits PAF-induced platelet aggregation
- Suitable for various research applications