Learn More
AdipoGen Forskolin,Chemical. CAS: 66428-89-5 and 66575-29-9
SDP
Supplier:  AdipoGen AGCN20089M050
Encompass_Preferred
Forskolin, AdipoGen Life Sciences. Chemical. CAS: 66428-89-5 and 66575-29-9. Formula: C22H34O7. MW: 410.5. Isolated from Coleus forskohlii. Potent, cell permeable adenylyl cyclase activator. Increases intracellular cAMP levels. Widely used tool to investigate cAMP as a second messenger. Inotropic and antihypertensive. Smooth muscle relaxant/vasodilator. Glucose transporter inhibitor. Platelet aggregation inhibitor. Stimulates lipolysis in fat cells. Non-competitive nicotinic acetylcholine receptors inhibitor. MAP kinase inhibitor. Upregulates mitochondrial uncoupling protein (UCP) mRNA levels in brown adipose tissue. Autophagy inhibitor. Hedgehog signaling inhibitor. Has antiglaucoma potential. Promotes neuronal differentiation of NSCs.
Catalog No. 50-596-450