Learn More
Medchemexpress LLC HY-N0028 5mg Medchemexpress, Forsythiaside A CAS:79916-77-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN00285MG
Encompass_Preferred
Medchemexpress, HY-N0028 5mg Forsythiaside A CAS:79916-77-1 Forsythiaside A, a phenylethanoside product isolated from air-dried fruits of Forsythia suspense, has anti-inflammatory and antioxidant effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0215