Learn More
Medchemexpress LLC Forsythoside E | 93675-88-8 | 98.6% | 462.45 | C20H30O12 | 10 MG

Supplier: Medchemexpress LLC HYN217310MG
Forsythoside E is a phenylethanoid glycoside isolated from the fruits of Forsythia suspensa (Thunb.) Vahl. It is provided as a research-grade natural product for laboratory studies and biochemical assays.
- High purity (98.6%).
- Molecular weight 462.45.
- Molecular formula C20H30O12.
- Available in small package sizes (5 mg, 10 mg, 50 mg, 100 mg).
- Suitable for in vitro biochemical and pharmacological studies.
- For research use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.