Learn More
Mac-Mod Fortis HILIC Diol, 100 A, 5 um, 2.1 x 100 mm HPLC Column
SDP
Supplier:  Mac-Mod FDI020505
Encompass
Fortis HILIC HPLC and UHPLC columns are packed with 100 A high-surface area silica-based columns designed with optimized bonding chemistries for uniform ligand coverage. Fortis HILIC HILIC and DIOL phases are available in a wide range of column IDs and column lengths including 75 micron, 200 micron, 2.1, 3.0, 4.6, 10 and 21.2 mm ID columns other column IDs and lengths available upon request. Guard columns are available and require guard holder DCGUA1.
Catalog No. 50-234-8111