Learn More
Mac-Mod Fortis SpeedCore-BIO Protein C4, 300 A, 3.5 um, 2.1 x 150 mm Core Shell Column
SDP
Supplier:  Mac-Mod SCPRO04020735
Encompass
Fortis SpeedCore BIO Core-Shell columns are designed with a smaller shell thickness that improves mass transfer of peptides and proteins. Fortis SpeedCore BIO columns are available in 160 A pore size materials for separations of peptides up to 15 kDa and 300 A pore sizes designed for Protein, mAb and other large biomolecule separations. Fortis SpeedCore BIO 160 A material is available in a 2.5 micron particle size and in analytical formats, in a C18 bonded phase chemistry. and Fortis SpeedCore BIO 300 A material is available in a C18, C8 and C4 bonded phase in a range of analytical formats.
Catalog No. 50-234-6969