Learn More
Selleck Chemical LLC Fosamprenavir calcium salt
SDP
Supplier:  Selleck Chemical LLC S65812MG
Encompass_Preferred
Fosamprenavir(GW433908G) is a pro-drug of the protease inhibitor and antiretroviral drug amprenavir It is used for the treatment of HIV-1 infections
Catalog No. 50-000-02079