Learn More
TARGETMOL CHEMICALS INC FOSAPREPITANT DIMEGLUMIN 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T179025MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fosaprepitant dimeglumine (MK-0517) is the dimeglumine salt form of fosaprepitant the water-soluble N-phosphorylated prodrug of aprepitant with antiemetic activity. Upon intravenous administration and rapid conversion to aprepitant this agent binds selectively to the human substance P/neurokinin 1 (NK1) receptors in the central nervous system (CNS). This inhibits receptor binding of the endogenous substance P and prevents substance P-induced emesis. purity: 98%
Catalog No. 50-257-7303